Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylsulfoxide, 99.9%, MilliporeSigma™

Catalog No. 50947470 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-470 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-470 Supplier MilliporeSigma Supplier No. MX1456NP20
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-68-5
Molecular Weight (g/mol) 78.14

Specifications

Melting Point 18.5°C
Quantity 20 L
Flash Point 87°C
Packaging Plastic Drum
Residue after Evaporation 5 ppm max
Assay Percent Range 99.9% min
Vapor Pressure 0.6 hPa (20°C)
Boiling Point 189°C (1013 hPa)
Solubility Information 1000 g/L
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.