Learn More
Medchemexpress LLC Methyltetrazine-acid | 1380500-88-8 | 99.75% | C11H10N4O2 | 100 MG
SDP
Catalog No. 5000356985
Encompass_Preferred
Methyltetrazine-acid is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-56985 Supplier Medchemexpress LLC Supplier No. HY141263100MG
Methyltetrazine-acid is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins