Learn More
Medchemexpress LLC Methyltetrazine-amido-PEG5-alkyne | 2322322-23-4 | C₂₄H₃₃N₃O₆ | 100 MG
SDP
Catalog No. 5000356028
Encompass_Preferred
Methyltetrazine-amido-PEG5-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other to a target protein, enabling selective degradation of target proteins through the intracellular ubiquitin-proteasome system. This product is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Purity and documentation available
Catalog No. 50-003-56028 Supplier Medchemexpress LLC Supplier No. HY141276100MG
Methyltetrazine-amido-PEG5-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand binds to an E3 ubiquitin ligase, and the other to a target protein, enabling selective degradation of target proteins through the intracellular ubiquitin-proteasome system. This product is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Purity and documentation available