Learn More
Medchemexpress LLC Methyltetrazine-amido-PEG5-alkyne | 2322322-23-4 | C24H33N5O6 | 50 MG
SDP
Catalog No. 5000353829
Encompass_Preferred
Methyltetrazine-amido-PEG5-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Targets E3 ubiquitin ligase and a protein
  • Utilizes ubiquitin-proteasome system for selective protein degradation
  • Compatible with click chemistry (alkynes, tetrazine)
  • Applicable in cancer and targeted cancer therapy research
Catalog No. 50-003-53829 Supplier Medchemexpress LLC Supplier No. HY14127650MG
Methyltetrazine-amido-PEG5-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Targets E3 ubiquitin ligase and a protein
  • Utilizes ubiquitin-proteasome system for selective protein degradation
  • Compatible with click chemistry (alkynes, tetrazine)
  • Applicable in cancer and targeted cancer therapy research