Learn More
Medchemexpress LLC Methyltetrazine-amine | 1345955-28-3 | MFCD28334555 | 99.4% | 201.23 | C10H11N5 | 5 MG
SDP
Catalog No. 5000223401
Encompass_Preferred
Methyltetrazine-amine is a tetrazine-based reagent used for site-specific dual functionalization of bioconjugates via inverse electron-demand Diels-Alder (iEDDA) click chemistry. It reacts selectively with strained alkenes to enable efficient bioconjugation, hydrogel crosslinking, and scaffold formation for tissue engineering and biomaterials research.
  • High purity suitable for bioconjugation (99.4%).
  • Reactive in inverse electron-demand Diels-Alder click reactions.
  • Solid, purple to purplish red appearance for easy handling.
  • Stable under recommended storage; protect from light.
  • Available in small-scale quantities for research use.
Catalog No. 50-002-23401 Supplier Medchemexpress LLC Supplier No. HY1351405MG
Methyltetrazine-amine is a tetrazine-based reagent used for site-specific dual functionalization of bioconjugates via inverse electron-demand Diels-Alder (iEDDA) click chemistry. It reacts selectively with strained alkenes to enable efficient bioconjugation, hydrogel crosslinking, and scaffold formation for tissue engineering and biomaterials research.
  • High purity suitable for bioconjugation (99.4%).
  • Reactive in inverse electron-demand Diels-Alder click reactions.
  • Solid, purple to purplish red appearance for easy handling.
  • Stable under recommended storage; protect from light.
  • Available in small-scale quantities for research use.