Learn More
Medchemexpress LLC Methyltetrazine-amine | 1345955-28-3 | MFCD28334555 | 99.4% | C10H11N5 | 10MG
SDP
Catalog No. 5000261817
Encompass_Preferred
Methyltetrazine-amine is a tetrazine-based reagent used as a bioorthogonal handle for site-specific dual functionalization of biomolecules. It is supplied as a pink to purplish-red solid (C10H11N5, MW 201.23) for research use, with high reported purity and storage guidance to protect from light.
  • Acts as a tetrazine bioorthogonal handle for rapid conjugation.
  • Enables site-specific dual functionalization of biomolecules.
  • Provided as a stable pink to purplish-red solid for easy handling.
  • High purity suitable for analytical and preparative applications.
  • Stable when stored protected from light; solvent storage at low temperatures recommended.
  • Soluble in common polar organic solvents such as DMSO and DMF.
Catalog No. 50-002-61817 Supplier Medchemexpress LLC Supplier No. HY13514010MG
Methyltetrazine-amine is a tetrazine-based reagent used as a bioorthogonal handle for site-specific dual functionalization of biomolecules. It is supplied as a pink to purplish-red solid (C10H11N5, MW 201.23) for research use, with high reported purity and storage guidance to protect from light.
  • Acts as a tetrazine bioorthogonal handle for rapid conjugation.
  • Enables site-specific dual functionalization of biomolecules.
  • Provided as a stable pink to purplish-red solid for easy handling.
  • High purity suitable for analytical and preparative applications.
  • Stable when stored protected from light; solvent storage at low temperatures recommended.
  • Soluble in common polar organic solvents such as DMSO and DMF.