Learn More
TARGETMOL CHEMICALS INC Methyltetrazine-Amine 25MG
SDP
Catalog No. 502877619
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Methyltetrazine-Amine is a methyl-substituted tetrazine amine used in the synthesis of PROTAC molecules. Purity 97.59%
Catalog No. 50-287-7619 Supplier TARGETMOL CHEMICALS INC Supplier No. T2096725MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Methyltetrazine-Amine is a methyl-substituted tetrazine amine used in the synthesis of PROTAC molecules. Purity 97.59%