Learn More
Medchemexpress LLC Methyltetrazine-amine hydrochloride | 1596117-29-1 | MFCD32061909 | 99.7% | 237.69 | C10H12ClN5 | 5 MG
SDP
Catalog No. 5000222509
Encompass_Preferred
Methyltetrazine-amine hydrochloride is a tetrazine-based reagent provided as the hydrochloride salt for bioorthogonal labeling and site-specific bioconjugation. It serves as a reactive tetrazine handle for inverse-electron-demand Diels-Alder (IEDDA) reactions with strained alkenes, enabling rapid, selective conjugation under mild conditions.
  • Provides a reactive tetrazine handle for IEDDA bioorthogonal chemistry.
  • Hydrochloride salt form improves solubility and handling.
  • High purity supports reliable conjugation and reproducible results.
  • Available in small milligram quantities for method development and optimization.
  • Suitable for site-specific dual functionalization of biomolecules.
Catalog No. 50-002-22509 Supplier Medchemexpress LLC Supplier No. HY135140A5MG
Methyltetrazine-amine hydrochloride is a tetrazine-based reagent provided as the hydrochloride salt for bioorthogonal labeling and site-specific bioconjugation. It serves as a reactive tetrazine handle for inverse-electron-demand Diels-Alder (IEDDA) reactions with strained alkenes, enabling rapid, selective conjugation under mild conditions.
  • Provides a reactive tetrazine handle for IEDDA bioorthogonal chemistry.
  • Hydrochloride salt form improves solubility and handling.
  • High purity supports reliable conjugation and reproducible results.
  • Available in small milligram quantities for method development and optimization.
  • Suitable for site-specific dual functionalization of biomolecules.