Learn More
Medchemexpress LLC Methyltetrazine-amine hydrochloride | 1596117-29-1 | MFCD32061909 | 99.7% | C10H12ClN5 | 10MG
SDP
Catalog No. 5000259260
Encompass_Preferred
Methyltetrazine-amine hydrochloride is a tetrazine-containing small-molecule reagent used for inverse electron demand Diels-Alder (iEDDA) bioorthogonal click chemistry. It facilitates rapid and selective conjugation to strained dienophiles for site-specific labeling and bioconjugation of biomolecules.
  • High chemical purity (99.7%) suitable for analytical and preparative workflows.
  • Hydrochloride salt form improves stability and handling as a solid.
  • Reactive tetrazine moiety enables fast, catalyst-free iEDDA reactions with dienophiles.
  • Suitable for site-specific biomolecule labeling and bioconjugation.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-59260 Supplier Medchemexpress LLC Supplier No. HY135140A10MG
Methyltetrazine-amine hydrochloride is a tetrazine-containing small-molecule reagent used for inverse electron demand Diels-Alder (iEDDA) bioorthogonal click chemistry. It facilitates rapid and selective conjugation to strained dienophiles for site-specific labeling and bioconjugation of biomolecules.
  • High chemical purity (99.7%) suitable for analytical and preparative workflows.
  • Hydrochloride salt form improves stability and handling as a solid.
  • Reactive tetrazine moiety enables fast, catalyst-free iEDDA reactions with dienophiles.
  • Suitable for site-specific biomolecule labeling and bioconjugation.
  • Available in small research pack sizes for laboratory use.