Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyltetrazine-DBCO | 1802238-48-7 | 95.2% | 683.73 | C34H33N7O7S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1403135MG

Encompass_Preferred

Methyltetrazine-DBCO is a heterobifunctional click chemistry reagent used as a non-cleavable antibody-drug conjugate (ADC) linker. It contains a DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC) with azides, and a methyltetrazine group that participates in inverse electron demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) partners.

  • Non-cleavable ADC linker suitable for bioconjugation.
  • Enables SPAAC reactions via the DBCO group.
  • Supports iEDDA reactions via the methyltetrazine moiety.
  • Molecular weight 683.73 and formula C34H33N7O7S.
  • Appearance: solid, purple to purplish red.
  • Purity approximately 95.2% (as supplied).
  • Available in small-scale packages, including 5 mg.
  • Storage: powder at -20°C (long term) or 4°C (shorter term); in solvent at -80°C or -20°C for limited durations.

Catalog No. 50-002-02679


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.