Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyltetrazine-Nhs Ester

Catalog No. 900914100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-091-4100MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-091-4100MG Supplier Sigma Aldrich Supplier No. 900914100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyltetrazine-NHS ester has an N-hydroxysuccinimide (NHS) ester that can be reacted with primary amines and a methyltetrazine, which is reactive to trans-cyclooctenes. The NHS ester reacts with primary amines (e.g. side chain of lysine residues or aminosilane-coated surfaces) at neutral or slightly basic pH to form covalent bonds. The short spacer arm adds minimal mass to modified molecules (230.2 daltons). It is generally used in click chemistry applications in bioconjugate chemistry.

Specifications

Melting Point 286°C to 287°C
Molecular Formula C24H39B3O6
Linear Formula C24H39B3O6
MDL Number MFCD17167305
Quantity 1 g
Synonym 1,3,5-Tris(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
Molecular Weight (g/mol) 456 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.