Learn More
Medchemexpress LLC Methyltetrazine-PEG24-NHS ester | 00-00-0 | 99.3% | C64H111N5O29 | 10MG
SDP
Catalog No. 5000261200
Encompass_Preferred
Methyltetrazine-PEG24-NHS ester is a PEG24 linker bearing a methyltetrazine reactive moiety and an N-hydroxysuccinimide (NHS) activated ester. Provided as a dry research reagent, it is designed for bioconjugation and PROTAC linker synthesis, enabling rapid bioorthogonal ligation to trans-cyclooctene (TCO) derivatives and stable amide bond formation with primary amines. The long PEG24 spacer enhances aqueous solubility and provides flexibility between conjugated components.
  • Enables rapid inverse electron-demand Diels-Alder (IEDDA) conjugation with TCO reagents.
  • Provides an NHS ester for efficient coupling to primary amines.
  • PEG24 spacer increases aqueous solubility and linker flexibility.
  • High purity suitable for research applications.
  • Supplied in mg-scale packages for small-scale conjugations.
  • Intended for research use only and not for human therapeutic use.
Catalog No. 50-002-61200 Supplier Medchemexpress LLC Supplier No. HY14127210MG
Methyltetrazine-PEG24-NHS ester is a PEG24 linker bearing a methyltetrazine reactive moiety and an N-hydroxysuccinimide (NHS) activated ester. Provided as a dry research reagent, it is designed for bioconjugation and PROTAC linker synthesis, enabling rapid bioorthogonal ligation to trans-cyclooctene (TCO) derivatives and stable amide bond formation with primary amines. The long PEG24 spacer enhances aqueous solubility and provides flexibility between conjugated components.
  • Enables rapid inverse electron-demand Diels-Alder (IEDDA) conjugation with TCO reagents.
  • Provides an NHS ester for efficient coupling to primary amines.
  • PEG24 spacer increases aqueous solubility and linker flexibility.
  • High purity suitable for research applications.
  • Supplied in mg-scale packages for small-scale conjugations.
  • Intended for research use only and not for human therapeutic use.