Learn More
Medchemexpress LLC Methyltetrazine-PEG24-NHS ester | 99.3% | C64H111N5O29 | 50 MG
SDP
Catalog No. 5000354022
Encompass_Preferred
Methyltetrazine-PEG24-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54022 Supplier Medchemexpress LLC Supplier No. HY14127250MG
Methyltetrazine-PEG24-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins