Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methyltetrazine-PEG4-acid | 1802907-91-0 | 99.8% | C20H28N4O7 | 25 MG
SDP

Catalog No. 5000357380
Encompass_Preferred

Methyltetrazine-PEG4-acid is a solid laboratory chemical primarily used for the manufacture of substances. It has a molecular weight of 436.46 and typically appears as a purple to purplish-red solid.

  • Purity: 99.8% (HPLC)
  • Molecular formula: C20H28N4O7
  • Stable under recommended storage conditions
  • Boiling point/range: 647.6±65.0 °C at 760 mmHg
  • Relative density: 1.236 g/cm³
  • Recommended storage:
    • Powder: -20°C for 3 years; 4°C for 2 years
    • In solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-57380 Supplier Medchemexpress LLC Supplier No. 25MGHY141264
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyltetrazine-PEG4-acid is a solid laboratory chemical primarily used for the manufacture of substances. It has a molecular weight of 436.46 and typically appears as a purple to purplish-red solid.

  • Purity: 99.8% (HPLC)
  • Molecular formula: C20H28N4O7
  • Stable under recommended storage conditions
  • Boiling point/range: 647.6±65.0 °C at 760 mmHg
  • Relative density: 1.236 g/cm³
  • Recommended storage:
    • Powder: -20°C for 3 years; 4°C for 2 years
    • In solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.