Learn More
CONJU PROBE LLC Methyltetrazine-PEG4-azide, 25mg | Chemical Formula: C21H30N8O5 | Molecular Weight: 474.51 | Purity: >95% by HPLC
Catalog No. NC3665976
Encompass
A heterobifunctional linker functionalized with a methyltetrazine moiety for inverse electron demand Diels-Alder cycloaddition reactions and an azide for click chemistry
Catalog No. NC3665976 Supplier CONJU PROBE LLC Supplier No. CP602225MG
A heterobifunctional linker functionalized with a methyltetrazine moiety for inverse electron demand Diels-Alder cycloaddition reactions and an azide for click chemistry