Learn More
Medchemexpress LLC Methyltetrazine-PEG4-maleimide | 1802908-02-6 | 98.6% | C24H30N6O7 | 10MG
SDP
Catalog No. 5000258704
Encompass_Preferred
Methyltetrazine-PEG4-maleimide is a PEG-based linker and click-chemistry reagent bearing a tetrazine moiety and a maleimide reactive group. Supplied as a powder for bioconjugation and PROTAC synthesis, it reacts via inverse electron-demand Diels-Alder (iEDDA) with trans-cyclooctene (TCO) partners.
Catalog No. 50-002-58704 Supplier Medchemexpress LLC Supplier No. HY14127810MG
Methyltetrazine-PEG4-maleimide is a PEG-based linker and click-chemistry reagent bearing a tetrazine moiety and a maleimide reactive group. Supplied as a powder for bioconjugation and PROTAC synthesis, it reacts via inverse electron-demand Diels-Alder (iEDDA) with trans-cyclooctene (TCO) partners.