Learn More
Medchemexpress LLC Methyltetrazine-PEG4-maleimide | 1802908-02-6 | 98.6% | C24H30N6O7 | 25 MG
SDP
Catalog No. 5000352629
Encompass_Preferred
Methyltetrazine-PEG4-maleimide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing a Tetrazine group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing TCO groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a Tetrazine group
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA) with TCO groups
Catalog No. 50-003-52629 Supplier Medchemexpress LLC Supplier No. 25MGHY141278
Methyltetrazine-PEG4-maleimide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing a Tetrazine group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing TCO groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a Tetrazine group
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA) with TCO groups