Learn More
CONJU PROBE LLC Methyltetrazine-PEG4-oxyamine, 25mg
Catalog No. NC3522328
Encompass
A heterobifunctional linker containing a methyltetrazine moiety for inverse electron demand Diels-Alder cycloaddition reactions and an oxyamine group for oxime ligation
Catalog No. NC3522328 Supplier CONJU PROBE LLC Supplier No. CP630625MG
A heterobifunctional linker containing a methyltetrazine moiety for inverse electron demand Diels-Alder cycloaddition reactions and an oxyamine group for oxime ligation