Learn More
Medchemexpress LLC Methyltetrazine-PEG5-alkyne | 1802907-97-6 | 98.0% | C20H26N4O5 | 100 MG
SDP
Catalog No. 5000356043
Encompass_Preferred
Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56043 Supplier Medchemexpress LLC Supplier No. HY141275100MG
Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins