Learn More
Medchemexpress LLC Methyltetrazine-PEG5-alkyne | 1802907-97-6 | 98.0% | C20H26N4O5 | 50 MG
SDP
Catalog No. 5000355316
Encompass_Preferred
Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker that facilitates the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are designed to harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound is provided as a pink to red solid with high purity, making it suitable for various research applications.
Key features and benefits:
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Supports selective degradation of target proteins
  • High purity of 98.0%
  • Good solubility in DMSO (≥ 100 mg/mL)
  • Stable when stored at 4°C under nitrogen
  • Extended stability in solvent at -80°C for 6 months, or -20°C for 1 month (under nitrogen)
Catalog No. 50-003-55316 Supplier Medchemexpress LLC Supplier No. HY14127550MG
Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker that facilitates the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are designed to harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound is provided as a pink to red solid with high purity, making it suitable for various research applications.
Key features and benefits:
  • PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Supports selective degradation of target proteins
  • High purity of 98.0%
  • Good solubility in DMSO (≥ 100 mg/mL)
  • Stable when stored at 4°C under nitrogen
  • Extended stability in solvent at -80°C for 6 months, or -20°C for 1 month (under nitrogen)