Learn More
Medchemexpress LLC Methyltetrazine-PEG5-methyltetrazine | 00-00-0 | 98.8% | 704.78 g/mol | C34H44N10O7 | 5 MG
SDP
Catalog No. 5000263034
Encompass_Preferred
Methyltetrazine-PEG5-methyltetrazine is a homobifunctional PEG-based linker bearing methyltetrazine groups at both termini. It is intended for bioorthogonal inverse electron demand Diels-Alder (iEDDA) click chemistry and as a linker in conjugation workflows; supplied as a solid for research use.
  • Homobifunctional methyltetrazine groups enable rapid iEDDA click reactions.
  • PEG5 spacer provides a flexible, hydrophilic linkage between reactive ends.
  • High purity (~98.8%) suitable for research applications.
  • Useful for PROTAC synthesis and biomolecule labeling workflows.
  • Purple to purplish red solid, easy to handle and weigh.
  • Soluble in DMSO; suggested concentration 100 mg/mL.
  • Store powder at -20 °C for long-term stability.
  • In solution, store at -80 °C for up to 6 months or -20 °C for up to 1 month.
  • For research use only; not for human or therapeutic use.
Catalog No. 50-002-63034 Supplier Medchemexpress LLC Supplier No. HY1334675MG
Methyltetrazine-PEG5-methyltetrazine is a homobifunctional PEG-based linker bearing methyltetrazine groups at both termini. It is intended for bioorthogonal inverse electron demand Diels-Alder (iEDDA) click chemistry and as a linker in conjugation workflows; supplied as a solid for research use.
  • Homobifunctional methyltetrazine groups enable rapid iEDDA click reactions.
  • PEG5 spacer provides a flexible, hydrophilic linkage between reactive ends.
  • High purity (~98.8%) suitable for research applications.
  • Useful for PROTAC synthesis and biomolecule labeling workflows.
  • Purple to purplish red solid, easy to handle and weigh.
  • Soluble in DMSO; suggested concentration 100 mg/mL.
  • Store powder at -20 °C for long-term stability.
  • In solution, store at -80 °C for up to 6 months or -20 °C for up to 1 month.
  • For research use only; not for human or therapeutic use.