Learn More
Medchemexpress LLC Methyltetrazine-PEG5-NHS ester | 1802907-92-1 | 97.0% | C24H31N5O9 | 100 MG
SDP
Catalog No. 5000356045
Encompass_Preferred
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules composed of two distinct ligands connected by a linker. These molecules harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Selectively degrades target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-56045 Supplier Medchemexpress LLC Supplier No. HY141269100MG
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules composed of two distinct ligands connected by a linker. These molecules harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Selectively degrades target proteins via the ubiquitin-proteasome system