Learn More
Medchemexpress LLC Methyltetrazine-PEG5-NHS ester | 1802907-92-1 | 97.0% | C24H31N5O9 | 25 MG
SDP
Catalog No. 5000352790
Encompass_Preferred
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system; they achieve this by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Degrades target proteins
  • Exploits intracellular ubiquitin-proteasome system
  • Connects a ligand for an E3 ubiquitin ligase with a ligand for the target protein
Catalog No. 50-003-52790 Supplier Medchemexpress LLC Supplier No. 25MGHY141269
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system; they achieve this by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Degrades target proteins
  • Exploits intracellular ubiquitin-proteasome system
  • Connects a ligand for an E3 ubiquitin ligase with a ligand for the target protein