Learn More
Medchemexpress LLC Methyltetrazine-PEG5-NHS ester | 1802907-92-1 | 97.0% | C24H31N5O9 | 50 MG
SDP
Catalog No. 5000353202
Encompass_Preferred
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Purity of 97.0%
Catalog No. 50-003-53202 Supplier Medchemexpress LLC Supplier No. HY14126950MG
Methyltetrazine-PEG5-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This product is for research use only.
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Purity of 97.0%