Learn More
Medchemexpress LLC Methyltetrazine-PEG8-acid | 2183440-33-5 | MFCD31580146 | 99.0% | C28H44N4O11 | 10MG
SDP
Catalog No. 5000259448
Encompass_Preferred
Methyltetrazine-PEG8-acid is a PEG8-based linker bearing a methyltetrazine moiety and a terminal carboxylic acid. It is formulated for bioorthogonal tetrazine ligation and conventional amide-coupling chemistries, enabling synthesis of PROTACs, bioconjugates, and labeled probes. The PEG8 spacer improves solubility and reduces steric interference in conjugation reactions.
  • Methyltetrazine warhead enables rapid, selective tetrazine ligation
  • PEG8 spacer improves aqueous solubility and reduces steric hindrance
  • Terminal carboxylic acid is compatible with standard amide coupling
  • High purity supports reproducible synthetic workflows
  • Supplied in small-scale quantities suitable for research use
Catalog No. 50-002-59448 Supplier Medchemexpress LLC Supplier No. HY14126510MG
Methyltetrazine-PEG8-acid is a PEG8-based linker bearing a methyltetrazine moiety and a terminal carboxylic acid. It is formulated for bioorthogonal tetrazine ligation and conventional amide-coupling chemistries, enabling synthesis of PROTACs, bioconjugates, and labeled probes. The PEG8 spacer improves solubility and reduces steric interference in conjugation reactions.
  • Methyltetrazine warhead enables rapid, selective tetrazine ligation
  • PEG8 spacer improves aqueous solubility and reduces steric hindrance
  • Terminal carboxylic acid is compatible with standard amide coupling
  • High purity supports reproducible synthetic workflows
  • Supplied in small-scale quantities suitable for research use