Learn More
Medchemexpress LLC Methyltetrazine-PEG8-NHS ester | 2183440-34-6 | MFCD31580145 | 99.3% | C32H47N5O13 | 10MG
SDP
Catalog No. 5000258926
Encompass_Preferred
Methyltetrazine-PEG8-NHS ester is a PEG-based linker bearing a methyltetrazine group and an N-hydroxysuccinimide (NHS) activated ester. It enables rapid bioorthogonal conjugation via inverse electron-demand Diels-Alder chemistry on the tetrazine side and efficient amine coupling through the NHS ester, making it suitable for PROTAC assembly and general bioconjugation in research applications.
  • Methyltetrazine reactive handle enables fast bioorthogonal cycloaddition.
  • NHS ester allows straightforward amine conjugation under mild conditions.
  • PEG8 spacer improves aqueous solubility and reduces steric hindrance.
  • High purity supports reliable synthetic and bioconjugation workflows.
  • Compatible with linker assembly and PROTAC synthesis workflows.
Catalog No. 50-002-58926 Supplier Medchemexpress LLC Supplier No. HY14127010MG
Methyltetrazine-PEG8-NHS ester is a PEG-based linker bearing a methyltetrazine group and an N-hydroxysuccinimide (NHS) activated ester. It enables rapid bioorthogonal conjugation via inverse electron-demand Diels-Alder chemistry on the tetrazine side and efficient amine coupling through the NHS ester, making it suitable for PROTAC assembly and general bioconjugation in research applications.
  • Methyltetrazine reactive handle enables fast bioorthogonal cycloaddition.
  • NHS ester allows straightforward amine conjugation under mild conditions.
  • PEG8 spacer improves aqueous solubility and reduces steric hindrance.
  • High purity supports reliable synthetic and bioconjugation workflows.
  • Compatible with linker assembly and PROTAC synthesis workflows.