Learn More
Medchemexpress LLC Methyltetrazine-Ph-NHS ester | 1644644-96-1 | 98.9% | 250 MG
SDP
Catalog No. 5000343028
Encompass_Preferred
Methyltetrazine-Ph-NHS ester is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. It acts as a click chemistry reagent, containing a Tetrazine group capable of undergoing an inverse electron demand Diels-Alder reaction (iEDDA) with molecules that have TCO groups.
  • Alkyl/ether-based protac linker
  • Utilized in the synthesis of protacs
  • Functions as a click chemistry reagent
  • Features a tetrazine group
  • Participates in an inverse electron demand Diels-Alder reaction (iEDDA)
  • Reacts with molecules containing TCO groups
Catalog No. 50-003-43028 Supplier Medchemexpress LLC Supplier No. 250MGHY130283
Methyltetrazine-Ph-NHS ester is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. It acts as a click chemistry reagent, containing a Tetrazine group capable of undergoing an inverse electron demand Diels-Alder reaction (iEDDA) with molecules that have TCO groups.
  • Alkyl/ether-based protac linker
  • Utilized in the synthesis of protacs
  • Functions as a click chemistry reagent
  • Features a tetrazine group
  • Participates in an inverse electron demand Diels-Alder reaction (iEDDA)
  • Reacts with molecules containing TCO groups