Learn More
Medchemexpress LLC Methyltetrazine-Ph-NHS ester | 1644644-96-1 | 98.9% | 5 G
SDP
Catalog No. 5000340666
Encompass_Preferred
Methyltetrazine-Ph-NHS ester is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing a Tetrazine group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing TCO groups.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a Tetrazine group that undergoes an inverse electron demand Diels-Alder reaction (iEDDA) with TCO groups
  • For research use only
Catalog No. 50-003-40666 Supplier Medchemexpress LLC Supplier No. HY1302835G
Methyltetrazine-Ph-NHS ester is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing a Tetrazine group that can undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing TCO groups.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a Tetrazine group that undergoes an inverse electron demand Diels-Alder reaction (iEDDA) with TCO groups
  • For research use only