Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methylthio)acetonitrile, 99%, Thermo Scientific™

Catalog No. AC171131000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171131000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171131000 Supplier Thermo Scientific Chemicals Supplier No. 171131000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35120-10-6
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00001925
InChI Key ZRIGDBVSVFSVLL-UHFFFAOYSA-N
Synonym methylthio acetonitrile, methylthioacetonitrile, methylthio-acetonitrile, 2-methylsulfanyl acetonitrile, acetonitrile, methylthio, ch3sch2cn, acmc-1afh2, methylsulfanyl acetonitrile #, inchi=1/c3h5ns/c1-5-3-2-4/h3h2,1h
PubChem CID 99295
IUPAC Name 2-methylsulfanylacetonitrile
SMILES CSCC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.03g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4800 to 1.4840 (20°C, 589nm)
Linear Formula CH3SCH2CN
Quantity 100 g
Fieser 08,349
Formula Weight 87.14
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material (Methylthio)acetonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.