Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylthio-DADMe-Immucillin A | 653592-04-2 | MFCD30738019 | 98.4% | 293.40 g/mol | C13H19N5OS | 100 MG
SDP

Catalog No. 5000249188
Encompass_Preferred

MT-DADMe-ImmA is a potent, selective inhibitor of human 5'-methylthioadenosine phosphorylase (MTAP) supplied as a white to off-white solid for laboratory research. It exhibits subnanomolar affinity (Ki ≈ 90 pM) and is supplied in milligram-scale packages appropriate for biochemical and cellular studies.

  • Potent MTAP inhibition with Ki ≈ 90 pM.
  • High purity (≈98.4%).
  • Molecular formula C13H19N5OS; molecular weight 293.40 g/mol.
  • White to off-white solid suitable for biochemical assays.
  • Available in small milligram package sizes for research use.
  • Recommended storage: powder -20°C for long-term stability.

Catalog No. 50-002-49188 Supplier Medchemexpress LLC Supplier No. HY101496100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MT-DADMe-ImmA is a potent, selective inhibitor of human 5'-methylthioadenosine phosphorylase (MTAP) supplied as a white to off-white solid for laboratory research. It exhibits subnanomolar affinity (Ki ≈ 90 pM) and is supplied in milligram-scale packages appropriate for biochemical and cellular studies.

  • Potent MTAP inhibition with Ki ≈ 90 pM.
  • High purity (≈98.4%).
  • Molecular formula C13H19N5OS; molecular weight 293.40 g/mol.
  • White to off-white solid suitable for biochemical assays.
  • Available in small milligram package sizes for research use.
  • Recommended storage: powder -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.