Learn More
Medchemexpress LLC HY-B0513 100mg Medchemexpress, Methylthiouracil CAS:56-04-2 Purity:>98%
SDP
Catalog No. 502082574
Encompass_Preferred
Medchemexpress, HY-B0513 100mg Methylthiouracil CAS:56-04-2 Methylthiouracil is an antithyroid agent. Methylthiouracil suppresses the production TNF-α and IL-6, and the activation of NF-κB and ERK1/2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2574 Supplier Medchemexpress LLC Supplier No. HYB0513100MG
Medchemexpress, HY-B0513 100mg Methylthiouracil CAS:56-04-2 Methylthiouracil is an antithyroid agent. Methylthiouracil suppresses the production TNF-α and IL-6, and the activation of NF-κB and ERK1/2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.