Learn More
TARGETMOL CHEMICALS INC Methylthiouracil 200MG
SDP
Catalog No. 502880734
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Methylthiouracil (NSC-193526) is a thiourea antithyroid agent that inhibits the synthesis of thyroid hormone, and it is used to treat hyperthyroidism. Purity 100%
Catalog No. 50-288-0734 Supplier TARGETMOL CHEMICALS INC Supplier No. T0304200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Methylthiouracil (NSC-193526) is a thiourea antithyroid agent that inhibits the synthesis of thyroid hormone, and it is used to treat hyperthyroidism. Purity 100%