Learn More
Hach Company Methylthymol Blue Indicator Powder Pillows, pk/100
Supplier:  Hach Company 2284799
Encompass
Indicator for metals and chelant titrations. Used in the Total Chelant Digital Titrator method, Hach method 8350
Catalog No. 50-199-5739