Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylthymol Blue, Pentasodium Salt 96%, Thermo Scientific™

Catalog No. AC414940100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC414940100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414940100 Supplier Thermo Scientific Chemicals Supplier No. 414940100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4310-80-9
CAS Min % 95.0
CAS Max % 100.0
Molecular Formula C37H44N2Na5O13S
InChI Key AEAATHGHTOCDOF-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C(=C2C=C(C(=O)C(=C2C)CN(CC(=O)O)CC(=O)O)C(C)C)C3=CC=CC=C3S(=O)(=O)O)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na].[Na]
Infrared Spectrum Authentic
IUPAC Name 2-[[5-[[3-[[bis(carboxymethyl)amino]methyl]-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene]-(2-sulfophenyl)methyl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
Molecular Weight (g/mol) 871.769
PubChem CID 131879314
Formula Weight 866.73
UV_Vis 95% min. (on dry substance)
Color Brown
Quantity 10g
Chemical Name or Material Methylthymol Blue, pentasodium salt, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.