Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Methylthymol blue, tetrasodium salt, pure, water soluble

Catalog No. AC106370050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106370050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106370050 Supplier Thermo Scientific Chemicals Supplier No. 106370050
  • $126.00
    $102.65 / Each of 1

    Save $23.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 130 min. min. (at lambda max, in water)
CAS 1945-77-3
Infrared Spectrum Authentic
Lambda Max 435 to 440nm (in water)
Loss on Drying 10% max. (110°C, 1 hr)
Packaging Glass bottle
Molecular Formula C37H44N2Na4O13S
MDL Number MFCD00148905
Beilstein 19, V,8, 619
Merck Index 15, 6202
Color Brown/Red to Green
Synonym 3,3'-Bis[N,N-di(carboxymethyl)aminomethyl]thymolsulfonephthalein, sodium salt, MTB
Solubility Information Solubility in water: 460g/L (20°C).
Quantity 5 g
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
Physical Form Powder
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
Molecular Weight (g/mol) 848.779
PubChem CID 131850582
Formula Weight 844.76
Grade Pure
Chemical Name or Material Methylthymol Blue, tetrasodium salt
Show More Show Less
EINECSNumber 217-743-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.