Learn More
Cayman Chemical Methyltransferase Fluorometric Assay Kit, 1 ea
SDP
Catalog No. 502896055
Encompass_Preferred
Cayman’s MT Fluorometric Assay is a continuous enzyme-coupled assay that can continuously monitor SAM-dependent MTs. The removal of the methyl group from SAM generates AdoHcy, which is rapidly converted to urate and H2O2 by an enzyme mixture provided in the kit. The reaction between H2O2 and ADHP (10-acetyl-3,7,-dihydroxyphenoxazine) produces the highly fluorescent compound resorufin, which is analyzed with an excitation wavelength between 530-540 nm and an emission wavelength between 585-595 nm. The assay is supplied with AdoHcy as a positive control. The assay can be used with any purified SAM-dependent MT.
Catalog No. 50-289-6055 Supplier Cayman Chemical Supplier No. 7001501 EA
Cayman’s MT Fluorometric Assay is a continuous enzyme-coupled assay that can continuously monitor SAM-dependent MTs. The removal of the methyl group from SAM generates AdoHcy, which is rapidly converted to urate and H2O2 by an enzyme mixture provided in the kit. The reaction between H2O2 and ADHP (10-acetyl-3,7,-dihydroxyphenoxazine) produces the highly fluorescent compound resorufin, which is analyzed with an excitation wavelength between 530-540 nm and an emission wavelength between 585-595 nm. The assay is supplied with AdoHcy as a positive control. The assay can be used with any purified SAM-dependent MT.