Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltri-n-butylammonium bromide, 98%, Thermo Scientific™

Catalog No. AAB2382830
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2382830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2382830 Supplier Thermo Scientific Chemicals Supplier No. B2382830

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37026-88-3
Molecular Formula C13H30BrN
Molecular Weight (g/mol) 280.29
MDL Number MFCD00043201
InChI Key FQUHTZCWXDFLBQ-UHFFFAOYSA-N
Synonym tributylmethylammonium bromide, methyltri-n-butylammonium bromide, 1-butanaminium, n,n-dibutyl-n-methyl-, bromide, methyltributylammonium bromide, methyl tributyl ammonium bromide, tributyl methyl azanium bromide, acmc-20ajfa, ksc493m4d, tributylmethylammonium bro-mide
PubChem CID 10062191
IUPAC Name tributyl(methyl)azanium;bromide
SMILES [Br-].CCCCC([NH3+])(CCCC)CCCC

Specifications

Melting Point 120°C to 122°C
Quantity 250 g
Beilstein 3704268
Sensitivity Hygroscopic
Formula Weight 280.29
Percent Purity 98%
Chemical Name or Material Methyltri-n-butylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-313-1
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.