Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyltrichlorosilane

Catalog No. M85301500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M85301500G 500 g
M85301100G 100 g
M853015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. M85301500G Supplier Sigma Aldrich Supplier No. M85301500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyltrichlorosilane (MTS) can be used:As a reagent for the surface-functionalization of fabrics and mesoporous materials to increase their hydrophobicity.As a source of silicon to synthesize silica doped carbon nanotubes.

Specifications

CAS 75-79-6
Density 1.273 g/mL at 25°C
Boiling Point 66°C
Molecular Formula CH3Cl3Si
Linear Formula CH3SiCl3
Refractive Index n20/D 1.411
MDL Number MFCD00000481
Quantity 500 g
Synonym MTS; Trichloro(methyl)silane
Molecular Weight (g/mol) 149.48 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.