Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyltrichlorosilane

Catalog No. M853015G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M853015G 5 g
M85301100G 100 g
M85301500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. M853015G Supplier Sigma Aldrich Supplier No. M853015G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyltrichlorosilane (MTS) can be used:As a reagent for the surface-functionalization of fabrics and mesoporous materials to increase their hydrophobicity.As a source of silicon to synthesize silica doped carbon nanotubes.

Specifications

CAS 138-09-0
Molecular Formula C4H4Na2O5
Refractive Index n20/D 1.411
Linear Formula C4H4O5Na2
MDL Number MFCD00064542
Quantity 5 g
Synonym MTS; Trichloro(methyl)silane
Molecular Weight (g/mol) 178.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.