Learn More
AdipoGen Methyltrioctadecylammonium Br
SDP
Catalog No. 502560521
Encompass_Preferred
Chemical. CAS 18262-86-7. Formula C55H114BrN. MW 869.42. Synthetic. Triple chain lipid. Long chain ionophore used in anion exchange for perchlorate-selective membranes. This is as sensing material with selectivity of chloride-selective polymeric membrane electrode. Organic surfactant. Used as alkylammonium cations for the design of layered chemical sensors.
Catalog No. 50-256-0521 Supplier AdipoGen Supplier No. CDXM0100G025
Chemical. CAS 18262-86-7. Formula C55H114BrN. MW 869.42. Synthetic. Triple chain lipid. Long chain ionophore used in anion exchange for perchlorate-selective membranes. This is as sensing material with selectivity of chloride-selective polymeric membrane electrode. Organic surfactant. Used as alkylammonium cations for the design of layered chemical sensors.