Learn More
Medchemexpress LLC Methyltrioctylammoni 100g
SDP
Supplier:  Medchemexpress LLC HYW099641100G
Encompass_Preferred
Methyltrioctylammonium hydrogen sulfate is a quaternary ammonium salt that is mainly used as an extraction solvent and a phase transfer catalyst in various chemical reactions It is also used as an electrolyte in electrochemical devices such as batteries and fuel cells and as a building block for the synthesis of pharmaceuticals and agrochemicals MTOAHS are multifunctional compounds with many potential industrial applications due to their reactivity stability and ability to selectively extract certain compounds from mixtures
Catalog No. 50-002-05700