Learn More
Strem, An Ascensus Company MTO, 98% 250mg
SDP
Catalog No. 5090113061
Encompass
CAS# 70197-13-6. 250mg. Methyltrioxorhenium(VII), 98%. MFCD00192332. Molecular Weight: 249.23. Molecular Formula: CH3ReO3. Color/form: colorless to pale-gray xtl. Strem# 75-2375. http://www.strem.com/catalog/v/75-2375/
Catalog No. 50-901-13061 Supplier Strem, An Ascensus Company Supplier No. 752375250MG
CAS# 70197-13-6. 250mg. Methyltrioxorhenium(VII), 98%. MFCD00192332. Molecular Weight: 249.23. Molecular Formula: CH3ReO3. Color/form: colorless to pale-gray xtl. Strem# 75-2375. http://www.strem.com/catalog/v/75-2375/