Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyltrioxorhenium(Vii)

Catalog No. 412910500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-291-0500MG 500 mg
41-291-0100MG 100 mg
41-291-02G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 41-291-0500MG Supplier Sigma Aldrich Supplier No. 412910500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyltrioxorhenium(Vii)

Specifications

CAS 70197-13-6
Melting Point 111°C
Molecular Formula CH3O3Re
Linear Formula CH3ReO3
MDL Number MFCD00192332
Quantity 500 mg
Synonym MTO; Methylrhenium(VII) trioxide
Molecular Weight (g/mol) 249.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.