Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriphenoxyphosphonium iodide, 95%, Thermo Scientific™

Catalog No. AC415020250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC415020250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC415020250 Supplier Thermo Scientific Chemicals Supplier No. 415020250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17579-99-6
Molecular Formula C19H18IO3P
Molecular Weight (g/mol) 452.228
InChI Key VKTOBGBZBCELGC-UHFFFAOYSA-M
Synonym methyltriphenoxyphosphonium iodide, methyltriphenoxyphosphanium iodide, acmc-20ake2, triphenyl phosphite methoiodide, methyl triphenoxyphosphonium iodide, methyl triphenoxy phosphanium iodide, methyltriphenoxyphosphorus 1+ iodide, phosphorus 1+ ,methyltriphenoxy-, iodide 1:1 , t-4
PubChem CID 2735090
IUPAC Name methyl(triphenoxy)phosphanium;iodide
SMILES C[P+](OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3.[I-]

Specifications

Melting Point 141.0°C to 146.0°C
Quantity 25 g
Solubility Information Solubility in water: negligible. Other solubilities: soluble in acetone, diethyl ether
Formula Weight 452.22
Chemical Name or Material Methyltriphenoxyphosphonium iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.