Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriphenylarsonium iodide, 98%, Thermo Scientific™

Catalog No. AAA1859914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1859914 25 g
AAA1859903 1 g
AAA1859906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1859914 Supplier Thermo Scientific Chemicals Supplier No. A1859914

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1499-33-8
Molecular Formula C19H18AsI
Molecular Weight (g/mol) 448.179
MDL Number MFCD00031543
InChI Key GHLKOMSNOFJXDT-UHFFFAOYSA-M
Synonym methyltriphenylarsonium iodide, arsonium, methyltriphenyl-, iodide, methyltriphenylarsanium iodide, methyl triphenylarsonium iodide, arsonium, methyltriphenyl-, iodide 1:1, c19h18as.i, acmc-209d3h, triphenylmethylarsonium iodide, methyl triphenyl arsanium iodide, arsonium,methyltriphenyl-, iodide 1:1
PubChem CID 73911
IUPAC Name methyl(triphenyl)arsanium;iodide
SMILES C[As+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[I-]

Specifications

Melting Point 173°C to 177°C
Quantity 25 g
UN Number UN3465
Beilstein 3639838
Sensitivity Light sensitive
Formula Weight 448.18
Percent Purity 98%
Chemical Name or Material Methyltriphenylarsonium iodide
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
EINECSNumber 216-108-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.