Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriphenylphosphonium Bromide, 98%

Catalog No. AC156950100
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156950100 10 g Plastic bottle
AC156951000 100 g Plastic bottle
AC156955000 500 g Plastic bottle
AC156950025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC156950100 Supplier Thermo Scientific Chemicals Supplier No. 156950100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1779-49-3 | C19H18BrP | 357.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1779-49-3
Molecular Formula C19H18BrP
Molecular Weight (g/mol) 357.23
MDL Number MFCD00011804
InChI Key LSEFCHWGJNHZNT-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
PubChem CID 74505
IUPAC Name methyl(triphenyl)phosphanium;bromide
SMILES [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 230.0°C to 234.0°C
Color White
Flash Point >240°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula CH3P(C6H5)3Br
Quantity 10 g
Fieser 01,697
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 950g/L methanol (20°C),soluble in aliphatic alcohols and chloroform,practically insoluble in arom. hydrocarbons and,acetone
Formula Weight 357.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyltriphenylphosphonium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 217-218-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.