Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyltriphenylphosphonium Chloride 98.0+%, TCI America™
SDP

Catalog No. M270225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M270225G 25 g
M27025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M270225G Supplier TCI America Supplier No. M270225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1031-15-8
Molecular Formula C19H18ClP
Molecular Weight (g/mol) 312.78
MDL Number MFCD00797851
InChI Key QRPRIOOKPZSVFN-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium chloride, methyl triphenyl phosphonium chloride, phosphonium, methyltriphenyl-, chloride, methyltriphenylphosphanium chloride, methyltriphenylphosphoniumchloride, methyl triphenylphosphoniumchloride, methyl triphenylphosphonium chloride, methyl triphenyl phosphanium chloride
PubChem CID 9879809
IUPAC Name methyltriphenylphosphanium chloride
SMILES [Cl-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 223°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 312.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyltriphenylphosphonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.