Learn More
Cayman Chemical Methyltrnsferase asy Buffe 1ea
SDP
Catalog No. 502892687
Encompass_Preferred
An ago-PAM of GLP-1R induces calcium mobilization in 9-3-H cells expressing human GLP-1R (EC50 1.3 UM) enhances GLP-1- exenatide- or liraglutide-induced calcium mobilization in the same cells (EC50s 1.8 8.4 and 30 UM respectively) potentiates exenatide-induced insulin secretion in primary mouse pancreatic islets at 30 UM reverses haloperidol-induced catalepsy in a rat model of Parkinson’s disease motor symptoms at 30 mg/kg
Catalog No. 50-289-2687 Supplier Cayman Chemical Supplier No. 7001411 EA
An ago-PAM of GLP-1R induces calcium mobilization in 9-3-H cells expressing human GLP-1R (EC50 1.3 UM) enhances GLP-1- exenatide- or liraglutide-induced calcium mobilization in the same cells (EC50s 1.8 8.4 and 30 UM respectively) potentiates exenatide-induced insulin secretion in primary mouse pancreatic islets at 30 UM reverses haloperidol-induced catalepsy in a rat model of Parkinson’s disease motor symptoms at 30 mg/kg