Learn More
Medchemexpress LLC α-Methyltyrosine methyl ester hydrochloride | 7361-31-1 | 98.9% | 245.71 | 10 G
SDP
Catalog No. 5000454746
Encompass_Preferred
α-Methyltyrosine methyl ester hydrochloride is an orally active and competitive tyrosine hydroxylase inhibitor. It can inhibit the conversion of tyrosine to dopamine. It causes kidney damage and urethral calculi in rats. It can be used as a tool for sympathetic nervous system research.
  • Orally active and competitive tyrosine hydroxylase inhibitor.
  • Inhibits the conversion of tyrosine to dopamine.
  • Causes kidney damage and urethral calculi in rats.
  • Can be used as a tool for sympathetic nervous system research.
Catalog No. 50-004-54746 Supplier Medchemexpress LLC Supplier No. HYW01340710G
α-Methyltyrosine methyl ester hydrochloride is an orally active and competitive tyrosine hydroxylase inhibitor. It can inhibit the conversion of tyrosine to dopamine. It causes kidney damage and urethral calculi in rats. It can be used as a tool for sympathetic nervous system research.
  • Orally active and competitive tyrosine hydroxylase inhibitor.
  • Inhibits the conversion of tyrosine to dopamine.
  • Causes kidney damage and urethral calculi in rats.
  • Can be used as a tool for sympathetic nervous system research.