Learn More
Cayman Chemical Methyllycaconitine (citrate), Purity: ≥98%, CAS: 351344-10-0, 10 mg
SDP
Catalog No. 502964444
Encompass_Preferred
An antagonist of α7-containing nAChRs (Ki = 1.4 nM)
Catalog No. 50-296-4444 Supplier Cayman Chemical Supplier No. 2139810 MG
An antagonist of α7-containing nAChRs (Ki = 1.4 nM)